Skip to Content
Merck
All Photos(2)

Key Documents

C144

Sigma-Aldrich

1-(3-Chlorophenyl)biguanide hydrochloride

solid

Synonym(s):

1-(m-Chlorophenyl)biguanide hydrochloride, N-(3-Chlorophenyl)imidodicarbonimidic diamide hydrochloride, m-CPBG hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NHC(NH)NHC(NH)NH2 · HCl
CAS Number:
Molecular Weight:
248.11
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

mp

190-194 °C (lit.)

solubility

H2O: soluble

SMILES string

Cl[H].NC(=N)NC(=N)Nc1cccc(Cl)c1

InChI

1S/C8H10ClN5.ClH/c9-5-2-1-3-6(4-5)13-8(12)14-7(10)11;/h1-4H,(H6,10,11,12,13,14);1H

InChI key

FOWAIJYHRWFTHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Very potent 5-HT3 serotonin receptor agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary A Rodd et al.
The Journal of pharmacology and experimental therapeutics, 321(3), 1003-1012 (2007-02-28)
Studies from our laboratory indicated that local perfusion of the ventral tegmental area (VTA) with a serotonin-3 (5-HT(3)) receptor agonist increased dopamine (DA) neuronal activity and that the self-infusion of ethanol (EtOH) and cocaine into the posterior VTA could be
Wen Liu et al.
Alcohol (Fayetteville, N.Y.), 40(3), 167-176 (2007-04-10)
The objectives of the present study were to (a) examine the effects of activating serotonin-3 (5-HT3) receptors on dopamine (DA) release in the anterior and posterior ventral tegmental area (VTA) of Wistar rats and (b) determine if there are differences
Jaime M Monti et al.
Progress in neuro-psychopharmacology & biological psychiatry, 35(5), 1341-1348 (2011-04-26)
The effects of the selective 5-HT(3) receptor agonist m-chlorophenylbiguanide (m-CPBG), and of the NMDA (N-methyl-D-aspartate) and AMPA (α-amino-3-hydroxy-5-methyl-4-isoxazole propionate)/kainate antagonists AP-5 [(±)-2-amino-5-phosphono-pentanoic acid] and CNQX (6-cyano-7-nitroquinoxaline-2,3-dione), respectively, were studied in adult male Wistar rats implanted for chronic sleep recordings. The
Philip M Lang et al.
Journal of neurophysiology, 96(6), 2963-2971 (2006-09-08)
Activity-dependent fluctuations in axonal excitability and changes in interspike intervals modify the conduction of trains of action potentials in unmyelinated peripheral nerve fibers. During inflammation of a nerve trunk, long stretches of axons are exposed to inflammatory mediators such as
G J Kilpatrick et al.
European journal of pharmacology, 182(1), 193-197 (1990-06-21)
1-(m-Chlorophenyl)-biguanide (mCPBG) was examined and compared with three 5-HT3 receptor agonists in three 5-HT3 receptor models. mCPBG inhibited [3H]GR67330 binding to 5-HT3 receptors with high affinity (IC50 1.5 nM). mCPBG depolarized the rat vagus nerve with an EC50 one tenth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service