
1-(3-Chlorophenyl)biguanide hydrochloride

Synonym(s):
1-(m-Chlorophenyl)biguanide hydrochloride, N-(3-Chlorophenyl)imidodicarbonimidic diamide hydrochloride, m-CPBG hydrochloride
Linear Formula:
ClC6H4NHC(NH)NHC(NH)NH2 · HCl
CAS Number:
Molecular Weight:
248.11
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Showing 1-1 of 1