Skip to Content
Merck
All Photos(1)

Key Documents

72516

Supelco

Bezafibrate

analytical standard

Synonym(s):

2-[4-[2-(4-Chlorobenzamido)ethyl]phenoxy]-2-methylpropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20ClNO4
CAS Number:
Molecular Weight:
361.82
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

loss

≤0.2% loss on drying

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1)C(O)=O

InChI

1S/C19H20ClNO4/c1-19(2,18(23)24)25-16-9-3-13(4-10-16)11-12-21-17(22)14-5-7-15(20)8-6-14/h3-10H,11-12H2,1-2H3,(H,21,22)(H,23,24)

InChI key

IIBYAHWJQTYFKB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

The peroxisome proliferator-activated receptor (PPAR) is a member of the steroid nuclear receptor superfamily. Bezafibrate is a peroxisome proliferator-activated receptor agonist for PPARα, PPARδ, and PPARγ. Lipoprotein lipase (LPL) activator.
PPARgamma agonists, including Bezafibrate, have beneficial effects in the suppression of the inflammatory response during RSV infection and therefore might have clinical efficacy in the course of severe RSV-infection.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Vasiliki Symeonidou et al.
Cell reports, 37(4), 109900-109900 (2021-10-28)
Infant MLL-AF4-driven acute lymphoblastic leukemia (ALL) is a devastating disease with dismal prognosis. A lack of understanding of the unique biology of this disease, particularly its prenatal origin, has hindered improvement of survival. We perform multiple RNA sequencing experiments on
Bezafibrate derivatives as potent effectors of hemoglobin.
C Poyart et al.
Methods in enzymology, 232, 496-513 (1994-01-01)
K L Goa et al.
Drugs, 52(5), 725-753 (1996-11-01)
The lipid-modifying profile of bezafibrate is characterised by marked decreases in elevated triglyceride levels, increases in high density lipoprotein (HDL) cholesterol levels and decreases in total and low density lipoprotein (LDL) cholesterol levels. Bezafibrate also reduces elevated levels of lipoprotein(a)
D Ståhlberg et al.
European journal of clinical pharmacology, 40 Suppl 1, S33-S36 (1991-01-01)
The influence of bezafibrate treatment on hepatic cholesterol metabolism was studied in rats and in humans. The activities of the three key enzymes involved in cholesterol metabolism [3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase, cholesterol 7 alpha-hydroxylase, and acyl-coenzyme A: cholesterol acyltransferase
E Kanterewicz et al.
Annals of the rheumatic diseases, 51(4), 536-538 (1992-04-01)
The case is presented of a 70 year old woman with mild hypercholesterolaemia and hypertension who was readmitted to hospital six months after a previous admission for angina pectoris. The patient was treated with verapamil, nifedipine, and aspirin, and had

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service