Skip to Content
Merck
All Photos(2)

Documents

64000H

Sigma-Aldrich

Midas® III-Plus Automated Stainer

suitable for hematology, histology, microbiology

Synonym(s):

Midas

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105202

IVD

for in vitro diagnostic use

Quality Level

application(s)

hematology
histology
microbiology

storage temp.

room temp

General description

Automated, benchtop stainer for hematology, histology, and microbiology
The Midas® III-Plus Automated Stainer gives you the ability to automate all hematology, histology, and microbiology staining applications. Since the unit is fully programmable, you can duplicate your existing staining techniques and ensure consistent results for your tissue, peripheral blood, and bone marrow smears, as well as your microbiological smears. We back up the Midas III-Plus Automated Stainer with a one year warranty and some of the best technical and customer service in the business. Don’t forget we have a full line of stains perfectly suited for use with the Midas III-Plus Automated Stainer.

Application

Automated, benchtop stainer for:


  • hematology
  • histology
  • microbiology

Features and Benefits

  • Utilizes the dip slide staining technique which ensures uniform staining for every sample. This reduces stain consumption up to 50%.
  • Ideally suited for Wright & Wright Giemsa staining of blood and bone marrow smears
  • Optional clear colorless enclosure with door
  • Convenient bench top size
  • Flexible programming to suit your protocols
  • Ensures sample integrity with the ability to connect to a laboratory purified water system, or Midas® Water Bath
  • Ideally suited for staining up to 300 slides per day

Legal Information

MIDAS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service