Skip to Content
Merck
All Photos(2)

Documents

64000WBF

Sigma-Aldrich

Midas® Water Bath Filters

for use with Midas® III-Plus Automated Stainer

Synonym(s):

filters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103706

compatibility

for use with Midas® III-Plus Automated Stainer

shipped in

ambient

General description

MIDAS®

Legal Information

MIDAS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service