Skip to Content
Merck
All Photos(2)

Key Documents

805505

Sigma-Aldrich

Poly(ethylene glycol) diglycidyl ether

Mn 1,000

Synonym(s):

Diepoxy PEG, PEG diglycidyl ether, Polyoxyethylene bis(glycidyl ether)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5O2-(C2H4O)n-C3H5O
CAS Number:
MDL number:
UNSPSC Code:
12162002
Pricing and availability is not currently available.

form

solid

mol wt

Mn 1,000

mp

35-40 °C

density

1.14 g/mL at 25 °C

storage temp.

2-8°C

InChI

1S/C8H14O4/c1(9-3-7-5-11-7)2-10-4-8-6-12-8/h7-8H,1-6H2

InChI key

AOBIOSPNXBMOAT-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Highlighting new synthetic modifications of PEG to improve the mechanical properties and degradation of resulting hydrogels in tissue engineering applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service