Skip to Content
Merck
All Photos(1)

Key Documents

177350

Sigma-Aldrich

1,2,3,4-Tetrahydro-1-naphthylamine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N · HCl
CAS Number:
Molecular Weight:
183.68
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

form

solid

mp

185-187 °C (lit.)

density

0.974 g/mL at 25 °C (lit.)

SMILES string

Cl.NC1CCCc2ccccc12

InChI

1S/C10H13N.ClH/c11-10-7-3-5-8-4-1-2-6-9(8)10;/h1-2,4,6,10H,3,5,7,11H2;1H

InChI key

DETWFIUAXSWCIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2,3,4-Tetrahydro-1-naphthylamine hydrochloride was used in enantiomeric separation using β-cyclodextrin-bonded negatively charged polyacrylamide gel-filled capillaries[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Koide et al.
Journal of chromatography. A, 893(1), 177-187 (2000-10-24)
Enantiomeric separation by capillary electrochromatography with beta-cyclodextrin-bonded negatively charged polyacrylamide gels was examined. The columns used are capillaries filled with a negatively charged polyacrylamide gel, a so-called monolithic stationary phase, to which allyl carbamoylated beta-CD (AC-beta-CD) derivatives covalently bind. The
J S Shin et al.
Biotechnology and bioengineering, 73(3), 179-187 (2001-03-21)
A kinetic resolution process for the production of chiral amines was developed using an enzyme-membrane reactor (EMR) and a hollow-fiber membrane contactor with (S)-specific omega-transaminases (omega-TA) from Vibrio fluvialis JS17 and Bacillus thuringiensis JS64. The substrate solution containing racemic amine
N S Kula et al.
Life sciences, 37(11), 1051-1057 (1985-09-16)
The optical isomers of apomorphine (APO) and N-propylnorapomorphine (NPA) were interacted with three biochemical indices of dopamine (DA) receptors in extrapyramidal and limbic preparations of rat brain tissue. There were consistent isomeric preferences for the R(-) configuration of both DA
Oxygen-18 studies on the oxidative deamination mechanism of alicyclic primary amines in rabbit liver microsomes.
H Kurebayashi et al.
Archives of biochemistry and biophysics, 215(2), 433-443 (1982-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service