Skip to Content
Merck
1,2,3,4-Tetrahydro-1-naphthylamine hydrochloride 98%

1,2,3,4-Tetrahydro-1-naphthylamine hydrochloride

1,2,3,4-Tetrahydro-1-naphthylamine hydrochloride 98%
Empirical Formula (Hill Notation):
C10H13N · HCl
CAS Number:
Molecular Weight:
183.68

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing