Saltar al contenido
MilliporeSigma
Todas las fotos(4)

Documentos clave

MABD168

Sigma-Aldrich

Anti-WNT1 Antibody, clone 10C8

ascites fluid, clone 10C8, from mouse

Sinónimos:

Proto-oncogene Wnt-1, Proto-oncogene Int-1 homolog

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

10C8, monoclonal

species reactivity

human, mouse

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WNT1(7471)

General description

Wnt-1, also known as Proto-oncogene Wnt-1 or Proto-oncogene Int-1 homolog, was first identified as a proto-oncogene activated by viral insertion in mouse mammary tumors. The Wnt-1 gene encodes a member of a large family of secreted proteins that are cysteine-rich, glycosylated, and poorly soluble. Wnt-1 serves as a ligand for the RYK coreceptor, resulting in Wnt signaling. The Wnt signalling pathway mediates the levels of cytosolic beta-catenin, a multi-functional protein that associates with membrane-bound E-cadherin as well as several DNA-binding proteins, such as members of the TCF/LEF family. Additionally, it serves a ligand for members of the frizzled family of seven transmembrane receptors and plays a role in development of the central nervous system.

Immunogen

Purified recombinant fragment of human WNT1 expressed in E. Coli.

Application

Detect Wnt-1 using this mouse monoclonal antibody, Anti-WNT1 Antibody, clone 10C8 validated for use in western blotting, Immunofluorescence, IHC & Flow Cytometry.
Immunofluorescence Analysis: A 1:200-1,000 dilution from a representative lot detected WNT1 in HeLa and 3T3-L1 cells.

Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected WNT1 in human adrenal tissue.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected WNT1 in HeLa cells.

Optimal working dilutions must be determined by end user.

Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in NIH/3T3, 3T3L1, and HeLa cell lysates.

Western Blot Analysis: A 1:500-2,000 dilution of this antibody detected WNT1 in NIH/3T3, 3T3L1, and HeLa cell lysates.

Target description

~41 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
NIH/3T3, 3T3L1, and HeLa cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

¿No encuentra el producto adecuado?  

Pruebe nuestro Herramienta de selección de productos.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico