Saltar al contenido
MilliporeSigma
Todas las fotos(1)

Documentos clave

268320

Millipore

Daptomycin, Streptomyces sp.

A semi-synthetic cyclic polypeptide antibiotic that is active against vancomycin-resistant gram-positive bacteria, including enterococci and Staphylococcus aureus (MIC₅₀ = 0.13 µg/ml).

Sinónimos:

Daptomycin, Streptomyces sp., Cubicin, Dapcin

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

Fórmula empírica (notación de Hill):
C72H101N17O26
Número de CAS:
Peso molecular:
1620.67
MDL number:
UNSPSC Code:
12352200

Quality Level

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to pale yellow

solubility

ethanol: 1 mg/mL (sonication may be required)
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C72H101N17O26/c1-5-6-7-8-9-10-11-22-53(93)81-44(25-38-31-76-42-20-15-13-17-39(38)42)66(108)84-45(27-52(75)92)67(109)86-48(30-59(102)103)68(110)89-61-37(4)115-72(114)49(26-51(91)40-18-12-14-19-41(40)74)87-71(113)60(35(2)24-56(96)97)88-69(111)50(34-90)82-55(95)32-77-63(105)46(28-57(98)99)83-62(104)36(3)79-65(107)47(29-58(100)101)85-64(106)43(21-16-23-73)80-54(94)33-78-70(61)112/h12-15,17-20,31,35-37,43-50,60-61,76,90H,5-11,16,21-30,32-34,73-74H2,1-4H3,(H2,75,92)(H,77,105)(H,78,112)(H,79,107)(H,80,94)(H,81,93)(H,82,95)(H,83,104)(H,84,108)(H,85,106)(H,86,109)(H,87,113)(H,88,111)(H,89,110)(H,96,97)(H,98,99)(H,100,101)(H,102,103)/t35?,36-,37?,43+,44+,45+,46+,47+,48+,49+,50-,60+,61?/m1/s1

InChI key

DOAKLVKFURWEDJ-OFNKPWESSA-N

General description

A semi-synthetic cyclic polypeptide antibiotic that is active against vancomycin-resistant gram-positive bacteria, including enterococci and Staphylococcus aureus (MIC50 = 0.13 µg/ml). Its bactericidal activities have been attributed to its role in disruption of membrane potential and increased efflux of potassium ions from bacterial cells, which lead to disruption of protein, DNA, and RNA synthesis. Exhibits superior bactericidal properties compared to vancomycin (Cat. No. 627850) even in the presence of serum.
A semi-synthetic cyclic polypeptide antibiotic that is active against vancomycin-resistant gram-positive bacteria, including enterococci and Staphylococcus aureus (MIC50 = 0.13 µg/ml). Its bactericidal activities have been attributed to its role in disruption of membrane potential and increased efflux of potassium ions from bacterial cells, which lead to disruption of protein, DNA, and RNA synthesis. Exhibits superior bactericidal properties compared to vancomycin (Cat. No. 627850) even in the presence of serum.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Kosmidis, C. and Levine, D.P., 2010. Expert Opin. Pharmacother.11, 615.
Cui, L., et al. 2006. Antimicrob. Agents Chemother.50, 1079.
Silverman, J.A., et al. 2003. Antimicrob. Agents Chemother.47, 2538.
Rybak, M.J., et al. 2000. Antimicrob. Agents Chemother.44, 1062.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Questions

Reviews

No rating value

Active Filters

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico