MilliporeSigma
  • The mammalian autophagy initiator complex contains 2 HORMA domain proteins.
The mammalian autophagy initiator complex contains 2 HORMA domain proteins.
Autophagy (2015-08-04)
Max Michel, Melanie Schwarten, Christina Decker, Luitgard Nagel-Steger, Dieter Willbold, Oliver H Weiergräber
ABSTRACT
ATG101 is an essential component of the ULK complex responsible for initiating cellular autophagy in mammalian cells; its 3-dimensional structure and molecular function, however, are currently unclear. Here we present the X-ray structure of human ATG101. The protein displays an open HORMA domain fold. Both structural properties and biophysical evidence indicate that ATG101 is locked in this conformation, in contrast to the prototypical HORMA domain protein MAD2. Moreover, we discuss a potential mode of dimerization with ATG13 as a fundamental aspect of ATG101 function.
Product Description
Supelco
Calcium-d-pantothenate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Poly(ethylene glycol), tested according to Ph. Eur., 400
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 3,350