MilliporeSigma
  • Horizontal Basal Cell-Specific Deletion of Pax6 Impedes Recovery of the Olfactory Neuroepithelium Following Severe Injury.
Horizontal Basal Cell-Specific Deletion of Pax6 Impedes Recovery of the Olfactory Neuroepithelium Following Severe Injury.
Stem cells and development (2015-03-27)
Jun Suzuki, Katsuyasu Sakurai, Maya Yamazaki, Manabu Abe, Hitoshi Inada, Kenji Sakimura, Yukio Katori, Noriko Osumi
ABSTRACT
In the mammalian olfactory epithelium (OE), olfactory receptor neurons (ORNs) are continuously regenerated throughout the animal's lifetime. Horizontal basal cells (HBCs) in the OE express the epithelial marker keratin 5 (K5) and the stem cell marker Pax6 and are considered relatively quiescent tissue stem cells in the OE. Pax6 is a key regulator of several developmental processes in the central nervous system and in sensory organs. Although Pax6 is expressed in the OE, its precise role remains unknown, particularly with respect to stem cell-like HBCs. To investigate the function of Pax6 in the developmental and regenerative processes in the OE, we generated conditional Pax6-knockout mice carrying a loxP-floxed Pax6 gene. Homozygous Pax6-floxed mice were crossed with K5-Cre transgenic mice to generate HBC-specific Pax6-knockout (Pax6-cKO) mice. We confirmed that the deletion of Pax6 expression in HBCs was sufficiently achieved in zone 1 of the OE in Pax6-cKO mice 3 days after methimazole-induced severe damage. In this condition, regeneration of the OE was dramatically impaired; both OE thickness and the number of ORNs were significantly decreased in the regenerated OE of Pax6-cKO mice. These results suggest that Pax6 expression is essential for HBCs to differentiate into neuronal cells during the regeneration process following severe injury.
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
Orotidine 5′-monophosphate trisodium salt, ≥99% (HPLC), powder
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline