MilliporeSigma
Method for the determination of chromium in feed matrix by HPLC.
Poultry science (2015-10-27)
Balakrishnan Umesh, Rajendra Moorthy Rajendran, Muthu Tamizh Manoharan
ABSTRACT
An improved method for the chromatographic separation and determination of chromium (III) and (VI) [ CRIII AND CRVI: ] in mineral mixtures and feed samples has been developed. The method uses precolumn derivatization using ammonium pyrrolidinedithiocarbamate ( APD: ) followed by reversed-phase liquid chromatography to separate the chromium ions. Both Cr(III) and Cr(VI) species are chelated with ammonium pyrrolidinedithiocarbamate prior to separation by mixing with acetonitrile and 0.5 mmol acetate buffer (pH 4.5). Optimum chromatographic separations were obtained with a polymer-based reversed-phase column (Kinetex, 5 μ, 250 × 4.5 mm, Phenomenex, Torrance, CA) and a mobile phase containing acetonitrile and water (7:3). Both Cr(III) and Cr(VI) ion concentrations were directly determined from the corresponding areas in the chromatogram. The effect of analytical parameters, including pH, concentration of ligand, incubation temperature, and mobile phase, was optimized for both chromium complexes. The range of the procedure was found to be linear for Cr(III) and Cr(VI) concentrations between 0.125 and 4 μg/mL (r² = 0.9926) and 0.1 and 3.0 μg/mL (r² = 0.9983), respectively. Precision was evaluated by replicate analysis in which the percentage relative standard deviation values for chromium complex were found to be below 4.0. The recoveries obtained (85-115%) for both Cr(III) and Cr(VI) complexes indicated the accuracy of the developed method. The degradation products, as well as the excipients, were well resolved from the chromium complex peak in the chromatogram. Finally, the new method proved to be suitable for routine analysis of Cr(III) and Cr(VI) species in raw materials, mineral mixtures, and feed samples.
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium dichromate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Potassium dichromate, BioXtra, ≥99.5%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Potassium dichromate, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Potassium dichromate, 99.98% trace metals basis
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ammonium pyrrolidinedithiocarbamate, ~99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N