MilliporeSigma
  • Identification of critical residues of linear B cell epitope on Goodpasture autoantigen.
Identification of critical residues of linear B cell epitope on Goodpasture autoantigen.
PloS one (2015-04-16)
Xiao-yu Jia, Zhao Cui, Jian-nan Li, Shui-yi Hu, Ming-hui Zhao
ABSTRACT
The autoantigen of anti-glomerular basement membrane (GBM) disease has been identified as the non-collagenous domain 1 of α3 chain of type IV collagen, α3(IV)NC1. Our previous study revealed a peptide on α3(IV)NC1 as a major linear epitope for B cells and potentially nephrogenic, designated as P14 (α3129-150). This peptide has also been proven to be the epitope of auto-reactive T cells in anti-GBM patients. This study was aimed to further characterize the critical motif of P14. 16 patients with anti-GBM disease and positive anti-P14 antibodies were enrolled. A set of truncated and alanine substituted peptides derived from P14 were synthesized. Circulating antibodies against the peptides were detected by enzyme linked immunosorbent assay (ELISA). We found that all sera with anti-P14 antibodies reacted with the 13-mer sequence in the C-terminus of P14 (P14c) exclusively. The level of antibodies against P14 was highly correlated with the level of antibodies against P14c (r=0.970, P<0.001). P14c was the core immunogenic region and the amino acid sequence (ISLWKGFSFIMFT) was highly hydrophobic. Each amino acid residue in P14c was sequentially replaced by alanine. Three residues of glycine142, phenylalanine143, and phenylalanine145 were identified crucial for antibody binding based on the remarkable decline (P<0.001) of antibody reaction after each residue replacement. We defined GFxF (α3142, 143,145) as the critical motif of P14. It may provide some clues for understanding the etiology of anti-GBM disease.
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available. Contact Technical Service
Supelco
Diethanolamine, analytical standard
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Diethanolamine, SAJ first grade, ≥98.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Diethanolamine, JIS special grade, ≥99.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Pricing and availability is not currently available. Contact Technical Service
Trolamine impurity B, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Pricing and availability is not currently available. Contact Technical Service
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Pricing and availability is not currently available. Contact Technical Service