MilliporeSigma
  • Biodegradable and biocompatible poly(ethylene glycol)-based hydrogel films for the regeneration of corneal endothelium.
Biodegradable and biocompatible poly(ethylene glycol)-based hydrogel films for the regeneration of corneal endothelium.
Advanced healthcare materials (2014-03-22)
Berkay Ozcelik, Karl D Brown, Anton Blencowe, Katharina Ladewig, Geoffrey W Stevens, Jean-Pierre Y Scheerlinck, Keren Abberton, Mark Daniell, Greg G Qiao
ABSTRACT
Corneal endothelial cells (CECs) are responsible for maintaining the transparency of the human cornea. Loss of CECs results in blindness, requiring corneal transplantation. In this study, fabrication of biocompatible and biodegradable poly(ethylene glycol) (PEG)-based hydrogel films (PHFs) for the regeneration and transplantation of CECs is described. The 50-μm thin hydrogel films have similar or greater tensile strengths to human corneal tissue. Light transmission studies reveal that the films are >98% optically transparent, while in vitro degradation studies demonstrate their biodegradation characteristics. Cell culture studies demonstrate the regeneration of sheep corneal endothelium on the PHFs. Although sheep CECs do not regenerate in vivo, these cells proliferate on the films with natural morphology and become 100% confluent within 7 d. Implantation of the PHFs into live sheep corneas demonstrates the robustness of the films for surgical purposes. Regular slit lamp examinations and histology of the cornea after 28 d following surgery reveal minimal inflammatory responses and no toxicity, indicating that the films are benign. The results of this study suggest that PHFs are excellent candidates as platforms for the regeneration and transplantation of CECs as a result of their favorable biocompatibility, degradability, mechanical, and optical properties.
Sigma-Aldrich
o-Dianisidine dihydrochloride, Suitable for use in glucose determination
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
o-Dianisidine dihydrochloride, ≥95%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Supelco
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile, ≥98%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Glucose (GO) Assay Kit, sufficient for 20 assays
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
L-Glutamine
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20