MilliporeSigma
  • SMAD2 and p38 signaling pathways act in concert to determine XY primordial germ cell fate in mice.
SMAD2 and p38 signaling pathways act in concert to determine XY primordial germ cell fate in mice.
Development (Cambridge, England) (2015-01-22)
Quan Wu, Kurumi Fukuda, Michael Weinstein, Jonathan M Graff, Yumiko Saga
ABSTRACT
The sex of primordial germ cells (PGCs) is determined in developing gonads on the basis of cues from somatic cells. In XY gonads, sex-determining region Y (SRY) triggers fibroblast growth factor 9 (FGF9) expression in somatic cells. FGF signaling, together with downstream nodal/activin signaling, promotes male differentiation in XY germ cells by suppressing retinoic acid (RA)-dependent meiotic entry and inducing male-specific genes. However, the mechanism by which nodal/activin signaling regulates XY PGC fate is unknown. We uncovered the roles of SMAD2/3 and p38 MAPK, the putative downstream factors of nodal/activin signaling, in PGC sexual fate decision. We found that conditional deletion of Smad2, but not Smad3, from XY PGCs led to a loss of male-specific gene expression. Moreover, suppression of RA signaling did not rescue male-specific gene expression in Smad2-mutant testes, indicating that SMAD2 signaling promotes male differentiation in a RA-independent manner. By contrast, we found that p38 signaling has an important role in the suppression of RA signaling. The Smad2 deletion did not disrupt the p38 signaling pathway even though Nodal expression was significantly reduced, suggesting that p38 was not regulated by nodal signaling in XY PGCs. Additionally, the inhibition of p38 signaling in the Smad2-mutant testes severely impeded XY PGC differentiation and induced meiosis. In conclusion, we propose a model in which p38 and SMAD2 signaling coordinate to determine the sexual fate of XY PGCs.
Sigma-Aldrich
Formamide, JIS special grade, ≥98.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formamide, SAJ first grade, ≥98.5%
SKU
Pack Size
Availability
Price
Quantity
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Trolamine, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Tretinoin, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetic anhydride, for GC derivatization, LiChropur, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Tamoxifen, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
4-Hydroxytamoxifen, (E) and (Z) isomers (50:50), analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, Arxada quality, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, JIS special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, JIS special grade, ≥98.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Triethanolamine, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetic anhydride