MilliporeSigma
  • Effect of inhaled MgSO4 on FEV1 and PEF in children with asthma induced by acetylcholine: a randomized controlled clinical trail of 330 cases.
Effect of inhaled MgSO4 on FEV1 and PEF in children with asthma induced by acetylcholine: a randomized controlled clinical trail of 330 cases.
Journal of tropical pediatrics (2013-12-18)
Y X Sun, C H Gong, S Liu, X P Yuan, L J Yin, L Yan, T T Shi, J H Dai
ABSTRACT
To determine the response of nebulized magnesium sulfate on the lung function of acetylcholine-induced asthma children. Three hundred and thirty children of asthma with positive bronchial provocation test were randomly divided into three groups: magnesium sulfate, albuterol, and a combination of magnesium sulfate and albuterol. Lung function was compared between the three groups. Forced expiratory volume in one second (FEV1) and peak expiratory flow (PEF) as percentage over predicted at 10 min and 20 min in albuterol and combination group were significantly improved when compared to magnesium group. The changes in FEV1 and PEF expressed as absolute and percentage over predicted was not statistically significant from baseline to 20 min in magnesium, albuterol, and combination of magnesium sulfate and albuterol. There was no significant adverse effect observed during the present study. Nebulized magnesium sulfate alone has a bronchodilatory effect in Ach-induced asthmatic children. The combination of MgSO4 and albuterol did not has a synergistic effect.
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Acetylcholine iodide, ≥97%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Salbutamol sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC
Supelco
Albuterol sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
Salbutamol
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Supelco
Salbutamol, VETRANAL®, analytical standard
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Salbutamol hemisulfate salt, ≥98%
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
USP
Acetylcholine chloride, United States Pharmacopeia (USP) Reference Standard
Salbutamol, European Pharmacopoeia (EP) Reference Standard
Acetylcholine chloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri