MilliporeSigma
  • Behavioural and pharmacological dissociation of chlordiazepoxide effects on discrimination and punished responding.
Behavioural and pharmacological dissociation of chlordiazepoxide effects on discrimination and punished responding.
Psychopharmacology (1986-01-01)
H Hodges, S Baum, P Taylor, S Green
ABSTRACT
Effects of chlordiazepoxide (CDP) and ethanolamine-O-sulphate (EOS) alone and in combination were tested on the acquisition and performance of continuous reinforcement - time out (CR-TO) and variable interval reinforcement - time out (VI-TO) operant discriminations in rats. CDP disrupted acquisition of CR-TO discrimination; effects were short lived, and neither CR-TO performance nor its reversal were impaired. Acquisition of VI-TO discrimination was increasingly impaired, and performance disrupted in pre-trained animals. EOS alone was inactive, and with CDP exerted only slight interactive effects. When a conflict component was added to the VI-TO schedule, however, EOS showed substantial additive effects with CDP on punished responding. The results suggest that CDP-induced increases in non-rewarded (TO) responding were related to task difficulty, pointing to a discrimination impairment rather than an anxiolytic effect. In addition, the specificity of EOS potentiation may reflect a pharmacological dissociation between CDP effects on discrimination and on punished responding, and suggest that GABA is selectively involved in resistance to punishment.
Sigma-Aldrich
2-Aminoethyl hydrogen sulfate, ≥98.0% (T)