Merck
  • Acrylic acid grafted guargum-nanosilica membranes for transdermal diclofenac delivery.
Acrylic acid grafted guargum-nanosilica membranes for transdermal diclofenac delivery.
Carbohydrate polymers (2012-11-06)
Arindam Giri, Tridib Bhunia, Samir Ranjan Mishra, Luna Goswami, Asit Baran Panda, Sagar Pal, Abhijit Bandyopadhyay
ABSTRACT
Green, hydrophobic device for controlled transdermal release of diclofenac sodium was designed from in situ nanosilica/acrylic acid grafted guargum membranes. Best grafting condition was assigned and nanocomposites were formed in situ using varying proportions of aqueous nanosilica sol. Nanocomposite/drug conjugates were formed by bringing down the medium pH from 9.0 to 7.0. The conjugates were characterized through infrared and solid state NMR spectroscopy, electron microscopy, hydro-swelling, surface contact angle, viscometry and biocompatibility. Most balanced property was exhibited by the membrane containing 1wt% nanosilica. It also had shown the highest encapsulation efficacy vis-à-vis slowest release as compared to others during experimentation in a Franz diffusion cell.
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Guar
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Zinc acrylate, 98%