MilliporeSigma
  • Enzymatically catalyzed DNA synthesis using L-Asp-dGMP, L-Asp-dCMP, and L-Asp-dTMP.
Enzymatically catalyzed DNA synthesis using L-Asp-dGMP, L-Asp-dCMP, and L-Asp-dTMP.
Chemistry & biodiversity (2008-01-22)
Montserrat Terrazas, Philippe Marlière, Piet Herdewijn
ABSTRACT
The replacement of the pyrophosphate moiety of deoxynucleoside triphosphates by L-aspartic acid allows incorporation of natural deoxynucleosides into DNA using HIV reverse transcriptase (RT) as enzyme, while retaining the canonical base-pair selectivity. N-Methylation of the L-aspartic acid leaving group results in a reduced fidelity of incorporation.
Sigma-Aldrich
2′-Deoxycytidine 5′-monophosphate, Sigma Grade, ≥95.0%