MilliporeSigma
  • Predictors of antimicrobial-resistant Escherichia coli in the feces of vegetarians and newly hospitalized adults in Minnesota and Wisconsin.
Predictors of antimicrobial-resistant Escherichia coli in the feces of vegetarians and newly hospitalized adults in Minnesota and Wisconsin.
The Journal of infectious diseases (2008-01-11)
Mark R Sannes, Edward A Belongia, Burney Kieke, Kirk Smith, Amy Kieke, Mary Vandermause, Jeff Bender, Connie Clabots, Patricia Winokur, James R Johnson
ABSTRACT
To determine whether poultry contact/consumption predicts colonization with antimicrobial-resistant Escherichia coli, 567 newly hospitalized patients and 100 vegetarians were assessed microbiologically and epidemiologically. Multivariable analysis showed that poultry contact/consumption, other dietary habits, and antimicrobial use did not significantly predict resistance. In contrast, foreign travel significantly predicted both trimethoprim-sulfamethoxazole resistance (prevalence ratio, 2.7 [95% confidence interval, 1.3-5.6]) and "any resistance" (total population), whereas intensive-care-unit exposure predicted any resistance (hospital patients). Thus, most of the individual-level exposures-including poultry contact/consumption-that had been expected to be significant risk factors for infection with antimicrobial-resistant E. coli did not prove to be such. Other exposures, including household-, community-, and population-level effects, may be more important.
Product Description
Supelco
Sulfamethizole, VETRANAL®, analytical standard
Supelco
Sulfamethizole, analytical standard, ≥99% (HPLC)