MilliporeSigma
  • Regulation of subtilase cytotoxin-induced cell death by an RNA-dependent protein kinase-like endoplasmic reticulum kinase-dependent proteasome pathway in HeLa cells.
Regulation of subtilase cytotoxin-induced cell death by an RNA-dependent protein kinase-like endoplasmic reticulum kinase-dependent proteasome pathway in HeLa cells.
Infection and immunity (2012-02-23)
Kinnosuke Yahiro, Hiroyasu Tsutsuki, Kohei Ogura, Sayaka Nagasawa, Joel Moss, Masatoshi Noda
ABSTRACT
Shiga-toxigenic Escherichia coli (STEC) produces subtilase cytotoxin (SubAB), which cleaves the molecular chaperone BiP in the endoplasmic reticulum (ER), leading to an ER stress response and then activation of apoptotic signaling pathways. Here, we show that an early event in SubAB-induced apoptosis in HeLa cells is mediated by RNA-dependent protein kinase (PKR)-like ER kinase (PERK), not activating transcription factor 6 (ATF6) or inositol-requiring enzyme 1(Ire1), two other ER stress sensors. PERK knockdown suppressed SubAB-induced eIF2α phosphorylation, activating transcription factor 4 (ATF4) expression, caspase activation, and cytotoxicity. Knockdown of eIF2α by small interfering RNA (siRNA) or inhibition of eIF2α dephosphorylation by Sal003 enhanced SubAB-induced caspase activation. Treatment with proteasome inhibitors (i.e., MG132 and lactacystin), but not a general caspase inhibitor (Z-VAD) or a lysosome inhibitor (chloroquine), suppressed SubAB-induced caspase activation and poly(ADP-ribose) polymerase (PARP) cleavage, suggesting that the ubiquitin-proteasome system controls events leading to caspase activation, i.e., Bax/Bak conformational changes, followed by cytochrome c release from mitochondria. Levels of ubiquitinated proteins in HeLa cells were significantly decreased by SubAB treatment. Further, in an early event, some antiapoptotic proteins, which normally turn over rapidly, have their synthesis inhibited, and show enhanced degradation via the proteasome, resulting in apoptosis. In PERK knockdown cells, SubAB-induced loss of ubiquitinated proteins was inhibited. Thus, SubAB-induced ER stress is caused by BiP cleavage, leading to PERK activation, not by accumulation of ubiquitinated proteins, which undergo PERK-dependent degradation via the ubiquitin-proteasome system.
Sigma-Aldrich
Sal003, ≥98% (HPLC)