MilliporeSigma
  • Characteristics and photo-responsive release property of liposome containing 7-acetoxy coumarin.
Characteristics and photo-responsive release property of liposome containing 7-acetoxy coumarin.
Journal of nanoscience and nanotechnology (2012-03-15)
Hee Jin Seo, Jin-Chul Kim
ABSTRACT
A photo-responsive liposome was developed by loading 7-acetoxy coumarin (ATC) in egg phosphatidylcholine (EPC) liposome. ATC was derivetized from 7-hydroxy coumarin using sodium acetate. The ATC-loaded liposomes were prepared by suspending the dry mixture film of ATC/EPC in distilled water and sonicating the suspension. When the ATC to EPC ratio was less than 1:8, homogeneous liposomal suspensions were obtained without any precipitate. On 1H NMR spectra, the unsaturated chains of liposomal EPCs were somewhat deteriorated by a subsequent irradiation, the irradiation of lambda = 365 nm and then the irradiation of lambda = 254 nm. On TEM photos, the size of liposomes incorporating ATC markedly increased by the subsequent irradiation. Photo-dimerization (under lambda = 365 nm) and de-dimerization (under lambda = 254 nm) of ATC took a place even in EPC liposomes. Upon the irradiation of lambda = 254 nm, liposome containing ATC dimers exhibited an enhanced release of 5(6)-carboxyfluorescein compared with liposome containing ATC monomer and liposome free of ATC. The de-dimerization of ATC dimers is believed to fluidize the liposomal membrane and promote the release.
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
5(6)-Carboxyfluorescein, suitable for fluorescence, BioReagent, ≥95% (HPLC)
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%