MilliporeSigma
  • Synthesis and pharmacology of 1-deoxy analogs of CP-47,497 and CP-55,940.
Synthesis and pharmacology of 1-deoxy analogs of CP-47,497 and CP-55,940.
Bioorganic & medicinal chemistry (2007-10-09)
John W Huffman, Alicia L S Thompson, Jenny L Wiley, Billy R Martin
ABSTRACT
A series of 1-deoxy analogs of CP-47,497 (8 and 13, n=0-7) and 1-deoxy analogs of CP-55,940 (9, n=0-7) have been synthesized and their affinities for the cannabinoid CB(1) and CB(2) receptors have been determined. Although the majority of these compounds exhibit selectivity for the CB(2) receptor, none have greater than modest affinity for either receptor. The interactions of these 1-deoxy nontraditional cannabinoids with the CB(2) receptor are discussed.
Product Description
Supelco
Δ9-Tetrahydrocannabinol solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
Sigma-Aldrich
Δ9-Tetrahydrocannabinol solution, ethanol solution
Sigma-Aldrich
1-Bromo-4-tert-butylbenzene, 97%