- 1-Bromoethene-1-sulfonyl fluoride (BESF) is another good connective hub for SuFEx click chemistry.
1-Bromoethene-1-sulfonyl fluoride (BESF) is another good connective hub for SuFEx click chemistry.
We demonstrate 1,2-dibromoethane-1-sulfonyl fluoride (DESF) as a bench-stable and readily accessible precursor to the robust SuFEx connector, 1-bromoethene-1-sulfonyl fluoride (BESF). The in situ generation of BESF from DESF opens up several new reaction profiles, including application in the syntheses of unprecedented 3-substituted isoxazole-5-sulfonyl fluorides, 1-substituted-1H-1,2,3-triazole-4-sulfonyl fluorides, 2-amino-1-bromoethane-1-sulfonyl fluorides and 4-bromo-β-sultams in good to excellent yields. These new modules comprise a pendant sulfonyl fluoride handle, which further undergoes facile and selective SuFEx reactions with a selection of aryl silyl ethers to generate stable and useful sulfonate connections.