MilliporeSigma
  • The product of a fos-related gene, fra-1, binds cooperatively to the AP-1 site with Jun: transcription factor AP-1 is comprised of multiple protein complexes.
The product of a fos-related gene, fra-1, binds cooperatively to the AP-1 site with Jun: transcription factor AP-1 is comprised of multiple protein complexes.
Genes & development (1989-02-01)
D R Cohen, P C Ferreira, R Gentz, B R Franza, T Curran
ABSTRACT
fra-1 encodes a serum-inducible protein (Fra-1) that is antigenically related to Fos. We have characterized Fra-1 expression in serum-stimulated cells using antibodies raised against several regions of this protein. Fra-1, expressed transiently in COS cells or in serum-stimulated rat fibroblasts, undergoes extensive post-translational modification, primarily by phosphorylation of serine residues. It is present in both the nucleus and the cytoplasm and participates in a protein complex with Jun. Using proteins synthesized in reticulocyte lysates, we have shown that Fra-1, like Fos, binds to the AP-1 recognition element cooperatively with Jun. A truncated Fra-1 protein that contains the leucine zipper region but not an adjacent basic amino acid domain, complexes with Jun in vitro but fails to bind AP-1 oligonucleotides. These results demonstrate that Fra-1 contributes to the DNA-binding activity ascribed to transcription factor AP-1.
Sigma-Aldrich
Fos-related antigen 1 human, recombinant, expressed in insect cells, ≥70% (SDS-PAGE)
Pricing and availability is not currently available.