MilliporeSigma
  • Sol-gel niobia sorbent with a positively charged octadecyl ligand providing enhanced enrichment of nucleotides and organophosphorus pesticides in capillary microextraction for online HPLC analysis.
Sol-gel niobia sorbent with a positively charged octadecyl ligand providing enhanced enrichment of nucleotides and organophosphorus pesticides in capillary microextraction for online HPLC analysis.
Journal of separation science (2017-12-28)
Sheshanka Kesani, Abdul Malik
ABSTRACT
A niobia-based sol-gel organic-inorganic hybrid sorbent carrying a positively charged C18 ligand (Nb2 O5 -C18 (+ve)) was synthesized to achieve enhanced enrichment capability in capillary microextraction of organophosphorus compounds (which include organophosphorus pesticides and nucleotides) before their online analysis by high-performance liquid chromatography. The sorbent was designed to simultaneously provide three different types of molecular level interactions: electrostatic, Lewis acid-base, and van der Waals interactions. To understand relative contributions of various molecular level analyte-sorbent interactions in the extraction process, two other sol-gel niobia sorbents were also created: (a) a purely inorganic sol-gel niobia sorbent (Nb2 O5 ) and (b) an organic-inorganic hybrid sol-gel niobia sorbent carrying an electrically neutral-bonded octadecyl ligand (Nb2 O5 -C18 ). The extraction efficiency of the created sol-gel niobia sorbent (Nb2 O5 -C18 (+ve)) was compared with that of analogously designed and synthesized titania-based sol-gel sorbent (TiO2 -C18 (+ve)), taking into consideration that titania-based sorbents present state-of-the-art extraction media for organophosphorus compounds. In capillary microextraction with high-performance liquid chromatography analysis, Nb2 O5 -C18 (+ve) had shown 40-50% higher specific extraction values (a measure of extraction efficiency) over that of TiO2 -C18 (+ve). Compared to TiO2 -C18 (+ve), Nb2 O5 -C18 (+ve) also provided superior analyte desorption efficiency (96 vs. 90%) during the online release of the extracted organophosphorus pesticides from the sorbent coating in the capillary microextraction capillary to the chromatographic column using reversed-phase high-performance liquid chromatography mobile phase.
Sigma-Aldrich
Trimethoxy(octadecyl)silane, technical grade
Pricing and availability is not currently available.
Sigma-Aldrich
2′-Deoxycytidine 5′-monophosphate, Sigma Grade, ≥95.0%
Pricing and availability is not currently available.