
Anti-Chloramphenicol Acetyltransferase Antibody, clone CAT-1

Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2