Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB3678

Sigma-Aldrich

Anti-Chloramphenicol Acetyltransferase Antibody, clone CAT-1

ascites fluid, clone CAT-1, Chemicon®

Synonym(s):

CAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

CAT-1, monoclonal

species reactivity

E. coli, vertebrates

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Chloramphenicol acetyl transferase (CAT). CAT is used as a tag in many systems and is also used in transcription assays.

Immunogen

Chloramphenicol acetyl transferase

Application

Detect Chloramphenicol Acetyltransferase using this Anti-Chloramphenicol Acetyltransferase Antibody, clone CAT-1 validated for use in WB & IC.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
Western blot: 1:2000-1:5000+

Immunocytochemistry: 4% PFA fixed cells (5-10′ RT, block with 3% normal secondary serum in PBS with 0.1% triton X-100 to permeabilize cell membrane. Primary antibody, 1:400, 1 hour.



Optimal working dilutions must be determined by the end user.

Physical form

Ascites fluid. Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CK2 inhibitors increase the sensitivity of HSV-1 to interferon-?.
Smith, MC; Bayless, AM; Goddard, ET; Davido, DJ
Antiviral research null
Benedikt M Markus et al.
mSphere, 4(3) (2019-06-28)
CRISPR-Cas9 technologies have enabled genome engineering in an unprecedented array of species, accelerating biological studies in both model and nonmodel systems. However, Cas9 can be inherently toxic, which has limited its use in some organisms. We previously described the serendipitous
Tânia Claro et al.
Microbiology (Reading, England), 159(Pt 1), 147-154 (2012-11-17)
Staphylococcus aureus is the major pathogen among the staphylococci and the most common cause of bone infections. These infections are mainly characterized by bone destruction and inflammation, and are often debilitating and very difficult to treat. Previously we demonstrated that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service