Key Documents
SYS-A310
WPI
WPI Single Channel Pulse Generator
Single channel pulse generator with train capability, TTL and variable voltage output
Select a Size
1 EA
$3,602.00
$3,602.00
Available to ship onMarch 11, 2025Details
Select a Size
Change View
1 EA
$3,602.00
About This Item
PDF Icon
A Trusted Marketplace Seller
PDF Icon
Have a question?
$3,602.00
Available to ship onMarch 11, 2025Details
General description
The A310 pulse generator/stimulator combines the reproducibility and accuracy of digital electronics with the fine resolution and continuous adjustment pos­si­ble with analog circuitry. All timing pa­ram­e­ters are entered with high resolution, ten-turn po­ten­ti­om­e­ters and six-position range switches. Timing is ac­cu­rate to within 1% of the set value.
Choose from one of the most recent versions:
Certificates of Analysis (COA)