Key Documents
503312
WPI
WPI Tissue Forceps
14.5cm, 3x4 Teeth
Select a Size
1 EA
$32.00
$32.00
Available to ship onMarch 11, 2025Details
Select a Size
Change View
1 EA
$32.00
About This Item
PDF Icon
A Trusted Marketplace Seller
PDF Icon
Have a question?
$32.00
Available to ship onMarch 11, 2025Details
General description
Tissue forceps with teeth have a fine tip with teeth for delicate tissue manipulation, such as during ophthalmic surgery or plastic surgery. Laboratory researchers choose these tissue forceps for precision work and microdissection. Choose between a 2x3, a 3x4, or a 4x5 tooth configuration. For example, 2x3 configuration has two teeth on one tip and three teeth on the other tip of the tweezers. The teeth ensure a firm grasp of the tissue, which is ideal for slippery or hard to hold tissues. While tweezers with teeth run the risk of puncturing delicate tissue, you do get a more secure hold. As a general rule, the fewer the teeth, the less likely the tissue trauma. These are available with straight tips. They are 14.5cm (5.7") long and fit easily into your hand for precision work. These are manufactured with surgical grade stainless steel, which is suitable for sterilization by autoclave or chemical sterilant. As with all fine surgical instruments, for best results and a long life for the instruments, always clean your surgical forceps thoroughly after each use.
Choose from one of the most recent versions:
Certificates of Analysis (COA)