Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900270

Sigma-Aldrich

Magnesium sulfate heptahydrate

Vetec, reagent grade, 99%

Synonym(s):

Epsom salts

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgSO4 · 7H2O
CAS Number:
Molecular Weight:
246.47
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

product line

Vetec

assay

99%

form

powder

pH

5-8 (50 g/L)

SMILES string

O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

Inchi Key

WRUGWIBCXHJTDG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E S Bain et al.
BJOG : an international journal of obstetrics and gynaecology, 121(5), 595-603 (2014-01-07)
To evaluate a slower (compared with a standard) infusion rate of the loading dose of magnesium sulphate for preterm fetal neuroprotection as a strategy to reduce maternal adverse effects. Randomised controlled trial. South Australian maternity hospital. Fifty-one women at <30
Oluwakemi Badaki-Makun et al.
Pediatric blood & cancer, 61(6), 1049-1054 (2014-01-21)
Multiple recent Sickle Cell Disease studies have been terminated due to poor enrollment. We developed methods to overcome past barriers and utilized these to study the efficacy and safety of intravenous magnesium for vaso-occlusive crisis (VOC). We describe the methods
N Arulkumaran et al.
Best practice & research. Clinical obstetrics & gynaecology, 27(6), 877-884 (2013-08-22)
Hypertensive disorders of pregnancy are one of the leading causes of peripartum morbidity and mortality globally. Hypertensive disease in pregnancy is associated with a spectrum of severity, ranging from mild pregnancy-induced hypertension to eclampsia. Although most cases of pre-eclampsia may
Chris Chin
Artificial organs, 37(1), 100-102 (2013-01-12)
Congenital heart disease (CHD) is relatively common and there are treatment options for virtually every form of CHD. With good care, the majority of babies can survive to adulthood; however, CHD remains a significant cause of death in infancy and
Karen J Gibbins et al.
Obstetrics and gynecology, 121(2 Pt 1), 235-240 (2013-01-25)
Clinical trials support the efficacy and safety of magnesium sulfate for cerebral palsy prevention. We evaluated the implementation of a clinical protocol for the use of magnesium for cerebral palsy prevention in our large women's hospital, focusing on uptake, indications

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service