Skip to Content
MilliporeSigma
All Photos(1)

Documents

V900115

Sigma-Aldrich

1,2-Propanediol

Vetec, reagent grade, 98%

Synonym(s):

Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein/REAXYS Number:
1340498
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

product line

Vetec

assay

98%

form

viscous liquid

autoignition temp.

779 °F

expl. lim.

12.5 %

refractive index

n20/D 1.432 (lit.)

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

217.4 °F - closed cup

flash_point_c

103 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R S Rajasenan et al.
Toxicology and applied pharmacology, 135(1), 89-99 (1995-11-01)
Engrafting components of human immune systems in severe combined immunodeficient (SCID) mice has been utilized to investigate the pathogenesis of several human autoimmune diseases and may provide a model for studying idiosyncratic drug toxicity. The purpose of this investigation was
Gai Ling Li et al.
Journal of controlled release : official journal of the Controlled Release Society, 84(1-2), 49-57 (2002-10-26)
To achieve a therapeutical effect of the anti-Parkinson's drug R-apomorphine via iontophoresis delivery, enhancement strategies in vitro were explored using three structurally related enhancers, lauric acid (LA), dodecyltrimethylammonium bromide (DTAB) and Laureth-3 oxyethylene ether (C(12)EO(3)). Human stratum corneum and shed
G A Reichard et al.
Diabetes, 35(6), 668-674 (1986-06-01)
Plasma acetone turnover rates were measured with the primed continuous infusion of 2-[14C]acetone in patients with moderate to severe diabetic ketoacidosis. Plasma acetone turnover rates ranged from 1.52 to 15.9 mumol X kg-1 X min-1 (108-1038 mumol X 1.73 m-2
H K Vaddi et al.
Journal of pharmaceutical sciences, 91(7), 1639-1651 (2002-07-13)
The respective alcoholic terpenes carvacrol, linalool, and alpha-terpineol were used at 5% w/v in propylene glycol (PG) to increase the in vitro permeation of haloperidol (HP) through human skin. The possible enhancement mechanism was then elucidated with HP-stratum corneum (SC)
Rong-Yao Wang et al.
Physical chemistry chemical physics : PCCP, 15(9), 3313-3319 (2013-01-31)
Understanding the role of kinetics in fiber network microstructure formation is of considerable importance in engineering gel materials to achieve their optimized performances/functionalities. In this work, we present a new approach for kinetic-structure analysis for fibrous gel materials. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service