Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800204

Sigma-Aldrich

Hydrochloric acid

0.1 N

Synonym(s):

Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Product Name

Hydrochloric acid solution, 0.1 N

vapor pressure

190 mmHg ( 25 °C)

product line

Vetec

form

liquid

concentration

0.1 N

bp

100 °C (lit.)

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrochloric acid is a strong acid, which is generally manufactured by dissolving HCl gas in water. It is largely used in ore processing, food manufacturing, steel pickling and in the production of calcium chloride. It′s a Bronsted acid used in the synthesis of amine salts and chlorides of benzyl, aryl, alkyl, vinyl, and allyl.

Application

Hydrochloric acid can be used:
  • In the study of corrosion behavior of aluminum 7075 alloy by electrochemical techniques.
  • Along with sodium nitrite for the diazotization of primary amines in the determination of abacavir sulfate and valacyclovir by spectrophotometric method.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical studies of aluminium 7075 alloy in different concentration of acid chloride medium
Pruthviraj RD and Rashmi M
Journal of Materials Science and Engineering. B, 5(221), 2169-0022 (2016)
A simple spectrophotometric determination of valacyclovir in pharmaceutical preparations
Srihari G, et al.
Journal of Current Chemical and Pharmaceutical Sciences, 1(3), 15-18 (2011)
Hydrochloric Acid
e-EROS Encyclopedia of Reagents for Organic Synthesis (2001)
Spectrophotometric methods for the determination of abacavir sulphate in pharmaceutical preparations
Srihari G
Global Journal of Pharmacology, 5(3), 172-175 (2011)
Marta Kolodziejczak et al.
ACS chemical neuroscience, 6(7), 1219-1230 (2015-04-11)
Maturation of functional neuronal circuits during central nervous system development relies on sophisticated mechanisms. First, axonal and dendritic growth should reach appropriate targets for correct synapse elaboration. Second, pruning and neuronal death are required to eliminate redundant or inappropriate neuronal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service