Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800008

Sigma-Aldrich

1-Hexanesulfonic acid sodium salt

suitable for HPLC, ≥99%

Synonym(s):

Sodium hexanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5SO3Na
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99%
Pricing and availability is not currently available.

product line

Vetec

assay

≥99%

technique(s)

HPLC: suitable

mp

>300 °C (lit.)

SMILES string

[Na+].CCCCCCS([O-])(=O)=O

InChI

1S/C6H14O3S.Na/c1-2-3-4-5-6-10(7,8)9;/h2-6H2,1H3,(H,7,8,9);/q;+1/p-1

InChI key

QWSZRRAAFHGKCH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philip Zakaria et al.
Electrophoresis, 23(17), 2821-2832 (2002-09-11)
The separation of a series of aromatic carboxylic acids, sulfonates and opiates using electrokinetic chromatography employing a mixture of the soluble cationic polymer poly(diallydimethylammonium chloride) (PDDAC) and the amphiphilic anion hexanesulfonate as pseudostationary phases is described. In this system, the
Kirti S Niralwad et al.
Ultrasonics sonochemistry, 17(5), 760-763 (2010-03-17)
1-Hexanesulphonic acid sodium salt was found to be an efficient catalyst for the green synthesis of alpha-aminophosphonates by the coupling of aldehydes/ketone, an amine and triethyl phosphite under ultrasound irradiation at ambient temperature for appropriate time to furnish the desired
A R Calhoun et al.
Journal of colloid and interface science, 309(2), 505-510 (2007-03-03)
Measurements have been made to determine the solubility of ethane, C2H6, in aqueous solutions of four different surfactants of the linear alkanesulfonate class at 25 degrees C. The surfactants, sodium 1-pentanesulfonate, sodium 1-hexanesulfonate, sodium 1-heptanesulfonate, and sodium 1-octanesulfonate, all share
R Salazar-González et al.
Molecular biology reports, 41(12), 7833-7843 (2014-08-29)
Arylamine N-acetyltransferase 2 (NAT2) metabolizes isoniazid (INH) and Single Nucleotide Polymorphisms (SNP) responsible for its activity has been reported. The aim of this study in the Mexican mestizo population was to evaluate NAT2 expression at the protein level in immune
Harshavardhan V Pawar et al.
Journal of pharmaceutical sciences, 103(6), 1720-1733 (2014-04-05)
Wafers combining weight ratios of Polyox with carrageenan (75/25) or sodium alginate (50/50) containing streptomycin and diclofenac were prepared to improve chronic wound healing. Gels were freeze-dried using a lyophilisation cycle incorporating an annealing step. Wafers were characterised for morphology

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service