Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V001741

Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine sulfate salt

98%

Synonym(s):

4-(Dimethylamino)aniline sulfate salt, 4-Amino-N,N-dimethylaniline sulfate salt, DMPPDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4NH2·H2SO4
CAS Number:
Molecular Weight:
234.27
Beilstein/REAXYS Number:
4612278
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

assay

98%

bp

495 °C (lit.)

mp

200-205 °C (dec.) (lit.)

SMILES string

OS(O)(=O)=O.CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2.H2O4S/c1-10(2)8-5-3-7(9)4-6-8;1-5(2,3)4/h3-6H,9H2,1-2H3;(H2,1,2,3,4)

InChI key

GLUKPDKNLKRLHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service