Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V001651

Supelco

Sodium 1-butanesulfonate

Synonym(s):

1-Butanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SO3Na
CAS Number:
Molecular Weight:
160.17
Beilstein/REAXYS Number:
3716662
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

mp

>300 °C (lit.)

SMILES string

[Na+].CCCCS([O-])(=O)=O

InChI

1S/C4H10O3S.Na/c1-2-3-4-8(5,6)7;/h2-4H2,1H3,(H,5,6,7);/q;+1/p-1

InChI key

XQCHMGAOAWZUPI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiro Narukawa et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 31(6), 521-527 (2015-06-13)
New measurement conditions for arsenic speciation analysis of rice flour were developed using HPLC-ICP-MS equipped with a reversed phase ODS column. Eight arsenic species, namely, arsenite [As(III)], arsenate [As(V)], monomethylarsonic acid (MMAA), dimethylarsinic acid (DMAA), trimethylarsine oxide (TMAO), tetramethylarsonium (TeMA)
Kristýna Šindelářová et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 32(12), 2027-2038 (2015-09-29)
A model small-scale field experiment was set up to investigate selenium (Se) uptake by four different varieties of broccoli plants, as well as the effect of Se foliar application on the uptake of essential elements for plants calcium (Ca), copper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service