Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V001450

Sigma-Aldrich

Bromophenol Red

Synonym(s):

3′,3″-Dibromophenolsulfonphthalein, Bromophenol red Sultone Form

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12Br2O5S
CAS Number:
Molecular Weight:
512.17
Beilstein/REAXYS Number:
354317
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

pH

5.2-6.8, yellow to red

SMILES string

Oc1ccc(cc1Br)C2(OS(=O)(=O)c3ccccc23)c4ccc(O)c(Br)c4

InChI

1S/C19H12Br2O5S/c20-14-9-11(5-7-16(14)22)19(12-6-8-17(23)15(21)10-12)13-3-1-2-4-18(13)27(24,25)26-19/h1-10,22-23H

InChI key

OYCLSQDXZMROJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service