Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V001293

Sigma-Aldrich

L-Rhamnose monohydrate

99%

Synonym(s):

6-Deoxy-L-mannose, L(+)Rhamnopyranose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5 · H2O
CAS Number:
Molecular Weight:
182.17
Beilstein/REAXYS Number:
3912643
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

assay

99%

SMILES string

O.C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H12O5.H2O/c1-2-3(7)4(8)5(9)6(10)11-2;/h2-10H,1H3;1H2/t2-,3-,4+,5+,6+;/m0./s1

InChI key

BNRKZHXOBMEUGK-NRBMBCGPSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivana R Sequeira et al.
Clinical and experimental pharmacology & physiology, 41(2), 107-117 (2013-09-17)
The effects of inflammatory changes on the absorption of different-sized probes and their permeability ratios are poorly understood. The aim of the present study was to determine the effects of a pharmacological agent on the permeability of the gut mucosa
James D Oliver et al.
Biotechnology and applied biochemistry, 62(3), 329-342 (2014-07-22)
Free-solution capillary electrophoresis (CE), or capillary zone electrophoresis, with direct UV detection was used for the first time for the determination of mono- and disaccharides, sugar alcohols, and ethanol in fermentation broths. Sample preparation proved to be minimal: no derivatization
Pavel Bartl et al.
International journal of food sciences and nutrition, 66(5), 514-519 (2015-06-20)
The anthocyanin composition of blue (Triticum aestivum L., cv. Skorpion) and purple wheat (Triticum aethiopicum JAKUBZ cv. Abyssinskaja arrasajta cv. Abyssinskaja arrasajta), cultivated in the Czech Republic, and of the prepared whole blue and purple wheat bread was determined. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service