Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V000602

Supelco

3-Hydroxy-4-nitroso-2,7-naphthalenedisulfonic acid disodium salt

p.a.

Synonym(s):

1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt, Nitroso-R salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5NNa2O8S2
CAS Number:
Molecular Weight:
377.26
Beilstein/REAXYS Number:
3643930
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

p.a.

SMILES string

[Na+].[Na+].Oc1c(N=O)c2ccc(cc2cc1S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C10H7NO8S2.2Na/c12-10-8(21(17,18)19)4-5-3-6(20(14,15)16)1-2-7(5)9(10)11-13;;/h1-4,12H,(H,14,15,16)(H,17,18,19);;/q;2*+1/p-2

InChI key

DMKMTGULLYISBH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Klara Midander et al.
Contact dermatitis, 70(6), 361-368 (2014-03-13)
Many daily contacts with metallic items are short and repetitive, and result in metal release; material, sweat, friction and wear may all be important. To study cobalt release and skin deposition as a result of many short and repetitive contacts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service