Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-2130

TriAltus™

TriAltus™ PreScission Protease

1 mg, Human Rhinovirus (HRV) 3C Protease overexpressed in E. coli with a glutathione S-transferase (GST) tag

Synonym(s):

PreScission, PreScission Protease, PSC, PSC P, Enzyme, Precission Enzyme, Protease

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By TriAltus Bioscience
A Trusted Marketplace Seller
PDF Icon
Have a question?

General description

The PreScission protease (PSC) is a human rhinovirus (HRV) type 14 3C protease fused to glutathione S-transferase (GST). This protease recognizes and cleaves the site specific sequence LEVLFQ/GP (where “/” indicates the cleavage site).


SPECIFICATIONS

PURITY | >99% (SDS gel analysis)

ENZYME SOURCE | Human Rhinovirus (HRV) 3C Protease overexpressed in E. coli with a glutathione S-transferase (GST) tag

CLEAVAGE SITE | PSC recognizes the sequence LEVLFQGP and cleaves between the glutamine (Q) and glycine (G)

CLEAVAGE EFFICIENCY | 20 µg protease will digest 2 mg protein with 99% efficiency at 4°C in 60 minutes.

STORAGE/SHIPPING CONCENTRATION | ~5 mg/mL

SHIPPING CONDITION | We ship PreScission Protease on dry ice in buffer (20 mM Tris (pH 8.0), 0.1 M NaCl, 1 mM EDTA, 50% glycerol, 1 mM βMe).

RECOMMENDED STORAGE CONDITIONS | -80 °C long term or -20 °C for 2-3 months in shipping buffer

EXPIRATION | 6 months from receipt when stored as directed


SKU | 

1 mg: 30-2130

5 mg: 30-2135

10 mg: 30-2140

50 mg: 30-2146

100 mg: 30-2150


Additional processing information: This highly efficient PreScission protease is compatible with TriAltus′ CLiM protein affinity purification technology.

Legal Information

Use of TriAltus products falls under our limited license agreement and is for non-profit research use only. For commercial use, please contact us for licensing.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library