Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SY3H94E1CBCB

Synthonix Corporation

n-Octadecylzinc bromide, 0.50 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
398.79
MDL number:
Product SY3H94E1CBCB is currently not available for purchase. Contact Technical Service

Iupac Name

bromo(octadecyl)zinc

Inchi Code

1S/C18H37.BrH.Zn/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2;;/h1,3-18H2,2H3;1H;/q;;+1/p-1

InChI key

JQNWQGPXQZOWEP-UHFFFAOYSA-M

storage temp.

Refrigerator

Purity

0.50

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solution

Product Link

https://synthonix.com/73289.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library