SY3H6E40EB14
Synthonix Corporation
(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(4-chloro-3-(4-ethoxybenzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate
About This Item
Iupac Name
(2R,3R,4R,5S,6S)-2-(acetoxymethyl)-6-(4-chloro-3-(4-ethoxybenzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate
Inchi Code
1S/C29H33ClO10/c1-6-35-23-10-7-20(8-11-23)13-22-14-21(9-12-24(22)30)26-28(38-18(4)33)29(39-19(5)34)27(37-17(3)32)25(40-26)15-36-16(2)31/h7-12,14,25-29H,6,13,15H2,1-5H3/t25-,26+,27-,28+,29+/m1/s1
InChI key
DKOQYKRDCDCNOR-ZCCUTQAASA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://synthonix.com/60628.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.