Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAAR4971600

Phenol red solution

pH indicator solution

Synonym(s):

Phenol Red, Phenolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14O5S
CAS Number:
Molecular Weight:
354.38
Beilstein/REAXYS Number:
326470
MDL number:
Pricing and availability is not currently available.

Product Name

Phenol Red Indicator Solution,

packaging

pkg of 100 mL (SAAR4971600JT)

visual transition interval

6.8-8.2, yellow to red

SMILES string

Oc1ccc(cc1)C2(OS(=O)(=O)c3ccccc23)c4ccc(O)cc4

InChI

1S/C19H14O5S/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19/h1-12,20-21H

InChI key

BELBBZDIHDAJOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For laboratory use only.
Suitable for use as a pH indicator.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service