Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90557

Supelco

4,4′-Thiobis(2-tert-butyl-5-methylphenol)

analytical standard

Synonym(s):

5-tert-Butyl-4-hydroxy-2-methylphenyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3CC6H2(CH3)OH]2S
CAS Number:
Molecular Weight:
358.54
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥99.0% (HPLC)

form

solid

shelf life

limited shelf life, expiry date on the label

mp

158-162 °C
163-165 °C (lit.)

SMILES string

Cc1cc(O)c(cc1Sc2cc(c(O)cc2C)C(C)(C)C)C(C)(C)C

InChI

1S/C22H30O2S/c1-13-9-17(23)15(21(3,4)5)11-19(13)25-20-12-16(22(6,7)8)18(24)10-14(20)2/h9-12,23-24H,1-8H3

InChI key

HXIQYSLFEXIOAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service