Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

698222

Liquid Level Indicator

Synonym(s):

Liquid Level Indicator with LED Readout Cube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111929
Pricing and availability is not currently available.

material

plastic packaging
stainless steel indicator

packaging

pkg of 1 unit

storage condition

room temperature

compatibility

for use with returnable containers

shipped in

ambient

General description

The Liquid Level Indicator with LED Readout Cube is a specialized solution designed for ReCycler® returnable containers, providing an efficient and precise method for monitoring chemical levels.This indicator serves as a valuable tool for industrial and scientific applications, ensuring accurate monitoring of chemical quantities while promoting efficiency and safety in handling returnable containers.

Legal Information

RECYCLER is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service