691127
QC Sampling Valve
Synonym(s):
QC Sampling Valve for ReCycler® returnable containers
About This Item
material
plastic packaging
stainless steel valve
packaging
pkg of 1 unit
storage condition
room temperature
compatibility
for use with returnable containers
shipped in
ambient
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service