Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691126

Jumper Hose Assembly

Synonym(s):

Jumper Hose Assembly for ReCycler® Bulk solvent containers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40142004
Pricing and availability is not currently available.

material

plastic packaging
stainless steel

packaging

pkg of 1 unit

storage condition

room temperature

compatibility

for use with returnable containers

shipped in

ambient

General description

The Jumper Hose Assembly is expertly engineered to provide a seamless, leak-free connection with ReCycler® Containers, ensuring exceptional reliability and durability in demanding industrial environments. It is a dependable solution for maintaining efficient and safe fluid transfer

Legal Information

RECYCLER is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service