Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691125

Ambient Air Vent

Synonym(s):

Ambient Air Vent for ReCycler® Bulk solvent containers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40101703
Pricing and availability is not currently available.

material

plastic packaging
stainless steel

packaging

pkg of 1 unit

storage condition

room temperature

compatibility

for use with returnable containers

shipped in

ambient

General description

Our ambient air vent, made from high-quality stainless steel, is specifically designed to integrate seamlessly with ReCycler® returnable bulk containers. Built to endure tough industrial conditions, it is also resistant to corrosion. This vent is essential as it helps balance the air pressure inside and outside the container. which is crucial for maintaining the stability and safety of the chemicals stored within.

Legal Information

RECYCLER is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service