Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

691122

Dispensing Hose Assembly

Synonym(s):

Dispensing hose with assembly for ReCycler® Bulk solvent Containers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40140000
Pricing and availability is not currently available.

material

plastic packaging
stainless steel

packaging

pkg of 1 unit

storage condition

room temperature

compatibility

for use with returnable containers

shipped in

ambient

General description

Dispensing Hose Assembly, specifically designed for use with ReCycler® returnable containers, offers a reliable solution to prevent chemical spills and efficiently extract chemicals from containers.
This innovative assembly ensures safe and controlled dispensing, minimizing the risk of accidental spills and promoting a cleaner, more efficient chemical handling process.

Legal Information

RECYCLER is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service